Learn More
Labcon North America 15mL PerformR Polystyrene Centrifuge Tubes, 125 per Bag
Supplier:  Labcon North America 31283500289
Encompass_Preferred
Tube, Conical centrifuge; Labcon; PerformR; 15mL; With attached flat screw caps, molded graduations, and writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Not Freezable; Not Autoclavable; Capacity: 15mL; Teal color caps; 125 tubes per bag/4 bags per case; 17mm W. 118mm H; Made in USA
Catalog No. 50-101-544