Learn More
Research Products International Corp Replacement Tube for Handheld UV Lamps, 6 Watt
SDP
Supplier:  Research Products International Corp 34001301
Encompass
Replacement Tube, 6 watt, for use with Handheld UV Lamps. Features UVC light (germicidal range of ultraviolet light). Wavelength is typically between 200 and 280 nm.
Catalog No. 50-136-7904