Learn More
Glass Expansion Tube Adaptor EZT4-16-1
SDP
Supplier:  Glass Expansion 708030697
Encompass_Preferred
Tube Adaptor EZT4-16-1. This tube adaptor fitting is used to connect this 1/16” OD capillary tube to a QSM-4 fitting.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3089