Learn More
Perkin Elmer US LLC Radial Purge Extension Window for Optima 5x00/7100/7200/7300 DV/8300
Supplier:  Perkin Elmer US LLC N0770944
Encompass_Preferred
The Radial View Purge Extension Window is a replaceable quartz window that slides into the mount between the torch and the optics for the radial view.
Catalog No. 50-904-3807