Learn More
Bellco Glass, Inc. TUBE & CAP, 50mL Centrifuge
SDP
Supplier:  Bellco Glass, Inc. 229456000
Encompass_Preferred
DescriptionGraduations are printed in bold, easy-to-read black ink and are accurate to +/-2%Large white marking area provides excellent contrast for identificationEasy to write on flat surface polyethylene (HDPE) caps feature integrated leak-proof sealTubes include graduations molded into the conical bottom, ideal for pellet viewing and small volume quantification Pack DescriptionTubes 500/Bag & Caps 500/BagCase Quantity500SterileNoAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: GreenPrimary MaterialTube: PolypropyleneSecondary MaterialCap: HDPEMaximum Volume50mLGraduatedYesGraduation Increments (Subdivisions)2.5mLMax Force (RCF)g16000
Catalog No. 50-233-0184