Learn More
TWD Scientific LLC Tube, Centrifuge,Twdbiox, 15Ml, Double Threaded, Flat Top, Sterile, Printed Graduations, Polypropylene, Conical Bottom, 500 Tubes/Case.
SDP
Supplier:  TWD Scientific LLC BX109151
Encompass_Preferred
BioX 15Ml Clear Polypropylene Sterile Conical Bottom Centrifuge Tubes With Printed Graduations. 25/Pack, Price/Case
Catalog No. 50-228-4980