Learn More
MSC Vikan Tube Brush, Hard, 40 mm Diameter, 500 mm Length, Orange
Supplier:  MSC 54670294
Encompass
This tube brush is designed for effective cleaning of bottles, tubes, and hard-to-reach surfaces. It features hard bristles, a 40 mm diameter, and a 500 mm length, making it suitable for demanding cleaning tasks.
  • Hard bristle stiffness for robust cleaning.
  • Constructed from durable polypropylene, polyester (PBT), and stainless steel (AISI 304Cu).
  • Designed for optimal performance in various cleaning environments.
  • Can be sterilized at up to 121 °C (autoclave) and cleaned at up to 93 °C (dishwasher).
  • Complies with food contact material regulations including (EC) 1935/2004, FDA CFR 21, and UK 2019 No. 704.
  • Manufactured according to EU Regulation 2023/2006/EC of Good Manufacturing Practice.
  • Free from intentionally added PFAS.
Catalog No. 50-283-5948