Learn More
Wilmad Labglass TUBE MELTING POINT 1.5X150MM
Supplier:  Wilmad Labglass LG9060106
Encompass
Tube, Melting Point, Capillary. Tube O.D.: 1.5mm; Tube O.D.: 1mm; Tube Length: 150mm. Fabricated from low extractable borosilicate glass. Capillary tube is closed at one end and packed in vials containing approximately 100 tubes.
Catalog No. 50-807-844
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.