Learn More
USA Scientific Inc 1.40 mL 2D Data-Matrix coded U-bottom push cap tube, bulk
Supplier:  USA Scientific Inc 17752607
Encompass
1.40 mL 2D Data-Matrix coded polypropylene push cap U-bottom tube (without cap), bulk, 960 tubes
Catalog No. NC0274156