Learn More
Chemglass Life Sciences Friction-Fit Tube Rack for 50 mL Centrifuge Tubes, Blue Polystyrene (Pack of 5)
SDP
Supplier:  Chemglass Life Sciences CLS4785050
Encompass
Friction-fit tube rack designed to hold 50 mL centrifuge tubes. Made from blue polystyrene for lightweight benchtop handling, sold as a pack of five. The friction-fit wells secure tubes for storage and short-term handling; not recommended for autoclaving.
  • Holds 50 mL centrifuge tubes.
  • Polystyrene construction for lightweight support.
  • Blue color for easy identification.
  • Friction-fit wells secure tubes without clamps.
  • Pack of 5 racks for multi-sample handling.
  • Not recommended for autoclaving.
Catalog No. NC2754184