Learn More
Saint Gobain Performance Plastics Tube Tygon 1/4 x 3/8 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAB00017
Encompass_Preferred
Saint-Gobain Tygon S3 Beverage Tubing Formula B-44-3 Overall Length 15.2 m 50 ft. Maximum Pressure 34 psi Diameter Inner 6.4 mm 1/4 in. Diameter Outer 9.5 mm 0.37 in. Autoclavable Autoclavable
Catalog No. 50-206-8913