Learn More
Accuflow Systems Tube, Tygn, 14 x38, 50ft/pk
Supplier:  Accuflow Systems ACF00017F
Encompass_Preferred
The Fisherbrand Tygon Tubing is a flexible, chemical-resistant PVC tubing that is 14mm in diameter with a 1.6mm wall thickness. It comes in a 50 foot pack and is autoclavable, with maximum temperatures up to 150°F. The tubing has a smooth inner wall for unrestricted flow and is non-toxic, non-hemolytic, and non-pyrogenic. It can be used for fluid transfer in analytical, clinical, and industrial applications. The tubing meets USP Class VI standards and is free of heavy metals, plasticizers, and latex.
Catalog No. 50-236-5761