Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, E-3603 PVC, 1/4 in ID, 50 ft
Supplier:  Accuflow Systems AAA00018
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 tubing is a flexible, non-DEHP PVC tubing engineered for resistance to flex-fatigue and abrasion. It is suitable for peristaltic pump lines, instrumentation connections, vents, drains, and general laboratory fluid transfer. Supplied in 50 ft continuous lengths, the tubing has a 1/4 in inner diameter and 7/16 in outer diameter.
  • Flexible PVC construction resists flex-fatigue and abrasion.
  • Phthalate-free formulation suitable for laboratory and food-contact use.
  • 1/4 in inner diameter, 7/16 in outer diameter, 3/32 in wall thickness.
  • 50 ft continuous length for extended routing without joins.
  • Clear wall allows visual monitoring of fluid flow.
  • Compatible with peristaltic pumps and common laboratory instrumentation.
Catalog No. 50-236-5922