Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Non-DEHP, 3/32 in ID, 5/32 in OD
Supplier:  Accuflow Systems AAA00004
Encompass_Preferred
Clear, flexible non-DEHP thermoplastic tubing designed for medical and laboratory fluid handling. This tubing provides biocompatible performance for blood-contact and peristaltic pump applications and tolerates common sterilization methods without significant loss of physical properties.
  • Biocompatible non-DEHP thermoplastic construction.
  • Inner diameter 3/32 in (2.38 mm), outer diameter 5/32 in (3.97 mm), wall thickness 1/32 in (0.79 mm).
  • Packaged length 50 ft.
  • Meets USP <88> and relevant ISO 10993 biocompatibility tests.
  • Compatible with autoclave, ethylene oxide, and gamma sterilization.
  • Clear and flexible for visual monitoring and peristaltic pumping.
  • Typical applications include blood and IV solutions, dialysis, and drug delivery.
Catalog No. 50-236-5801