Learn More
Saint Gobain Performance Plastics Tube Tygon 5/16 x 9/16 50FT/PK
Supplier:  Saint Gobain Performance Plastics AAA00024
Encompass_Preferred
Saint-Gobain Tygon S3 Clear Tubing Formula B-44-4X Diameter Inner 5/16 in. 7.9 mm Diameter Outer 0.56 in. 14 mm Autoclavable Autoclavable Certifications/Compliance FDA 21 CFR 175.300 3-A and NSF Standard 51 Hardness 65 (Shore A) Max. Operating Temperature 165 deg.F 79 deg.C Must order minimum quantity of 2.
Catalog No. 50-206-8889