Learn More
Creative Biomart Tuberculin purified protein derivative (PPD)
Supplier:  Creative Biomart THP0002
Encompass_Preferred
The product is a sterile aqueous solution of a purified protein fraction isolated from culture media filtrates of a human strain of Mycobacterium tuberculosis.
Catalog No. 50-208-4053