Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Platinum-Cured, 0.188 in ID, 0.375 in OD
Supplier:  Accuflow Systems STHTC1873H
Encompass_Preferred
Platinum-cured silicone tubing designed for sanitary fluid transfer and bioprocess applications. It features a smooth bore and flexible wall, balancing low extractables with resistance to repeated sterilization cycles. Supplied in 100 ft coils, the tubing is suitable for laboratory and process lines requiring biocompatible, autoclavable tubing.
  • Platinum-cured silicone material.
  • 0.188 in inner diameter, 0.375 in outer diameter, 0.093 in wall thickness.
  • 50 Shore A hardness for flexibility and dimensional stability.
  • Supplied in 100 ft coils for extended runs.
  • Autoclavable and gamma stable for repeated sterilization.
  • Meets USP Class VI and European Pharmacopoeia 3.1.9 standards.
  • Wide temperature range, -62°C to 260°C (-80°F to 500°F).
  • Smooth bore minimizes particulate trapping and flow disruption.
Catalog No. NC3233188