Learn More
Grainger Tubing, Nylon 1/4IN Yellow
Supplier:  Grainger 2VDV6
Encompass_Preferred
The Grainger 2VDV6 Nylon Tubing is a flexible nylon tubing with a 1/4" OD and a 1/8" ID. It has a maximum pressure rating of 850 psi at 73°F. The tubing is translucent so contents are visible and has a temperature range of -40°F to 160°F. It is sold by the foot and made in the USA. The nylon material offers good flex life and abrasion resistance. Common applications include air lines, water lines, and hydraulic fluids.
Catalog No. NC2745175