Learn More
Waters Corp Waters Tubing Assembly, U-Column Connection, 0.004 in. ID, UPLC
Supplier:  Waters Corp 700009535
Encompass_Preferred
This tubing assembly is a U-shaped column connector with a 0.004 in. internal diameter, specifically designed for use in UPLC systems. It is a vital service part for maintaining the functionality of components like the column manager and column heater within these systems.
  • Designed as a U-shaped column connector.
  • Features a 0.004 in. internal diameter.
  • Compatible with UPLC systems.
  • Suitable for use with column manager and column heater components.
  • Supplied as one unit per package.
  • Recommended for replacement as required.
Catalog No. 50-203-9669