Learn More
Waters Corp Waters Tubing, Clear, TFE, 1/16 in ID, 25 ft
Supplier:  Waters Corp WAT077043
Encompass_Preferred
This clear TFE tubing functions as a sample loop waste line, designed to allow solvent to flow efficiently into a waste container, making it suitable for HPLC system applications.
  • Material: TFE
  • Inner diameter: 1/16 in
  • Length: 25 ft
  • Suitable for high-performance liquid chromatography (HPLC) systems.
  • Functions as a sample loop waste line.
  • Designed to allow solvent to flow into a waste container.
Catalog No. 50-793-714