Learn More
Microsolv Technology Corporation Tubing Connector Fittings, High Pressure, One Piece, 1/16", Carbon PEEK, Black, Winged Head with 10-32 Screw Threads. Finger Tighten, 10/PK.
Supplier:  Microsolv Technology Corporation 4730110
Encompass_Preferred
These "Male Nuts" are used to connect PEEK or Stainless Steel tubing in the high pressures zones of HPLC systems. Fitting has a 15mm diameter knurled head that is "winged" for ease of generating torque, 21mm total length (top to bottom) with a 13mm thread length. Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
Catalog No. 50-227-1448