Learn More
Cole-Parmer Fitting, 304 Stainless Steel, Sanitary Clamp, 4"
Supplier:  Cole-Parmer 5011847
Encompass_Preferred
Use the sanitary clamps with sanitary fittings to create a leak-proof connection. These easy-to-use clamps are made from 304 stainless steel to provides good chemical resistance.
Catalog No. 50-255-7741