Learn More
Microsolv Technology Corporation Tubing Fep 1/16 X.040 5M
Supplier:  Microsolv Technology Corporation 4841040
Encompass_Preferred
Tubing, FEP, 0.040 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51609