Learn More
Waters Corp Tubing Kit, ACQUITY UPLC CM-A, Stainless Steel, 2-Column
Supplier: Waters Corp 205001819
This tubing kit comprises stainless steel tubing assemblies designed for integrating a CM-A active column manager with an ACQUITY H-Class PLUS system, enabling a two-column setup. It can also serve as a complete replacement for all stainless steel tubing in such a configuration, and it includes the necessary Active Preheater for a PLUS system.
- Type: Fitting, tool
- System type: UPLC
- Instrument compatibility: Arc Premier system, H-Class system, Patrol
- Replacement schedule: As required
- Component: QSM
- UNSPSC: 41115720