Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, BioPharm Platinum-Cured Silicone, L/S 13; 25 ft
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX9642013/DEL/DEL
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent