Learn More
Accuflow Systems Masterflex C-Flex spooled, L/S16, 400 ft.
Supplier:  Accuflow Systems 0642716
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. FDA compliant and pptically inspected and factory tested. Ensures that your tubing is clean, free from defects, and performs to exact specifications.Thirteen tubing formulations are available. Each formulation has different chemical compatibility properties, agency approvals, and performance characteristics. Norprene, PharMed BPT, and Tygon formulations are the firmest tubing we offer; silicone and C-Flex are the softest tubing. Firm tubing has longer life, better pressure generation, a stronger vacuum/suction lift, and lower gas permeability than softer tubings.
Catalog No. 50-110-8956