Learn More
Accuflow Systems TUBING MFLEX C-FLEX #90 10FT
Supplier:  Accuflow Systems 0642490
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex C-Flex tubing, I/P 90, 10 ft. Applications: General-purpose lab uses requiring high performance and long life. Characteristics: Exceptional tensile and tear strength; good biocompatibility; low
Catalog No. 50-110-9243