Learn More
Accuflow Systems TUBING MFLEX NORPRENE #16 50FT
Supplier:  Accuflow Systems 0640416
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Norprene tubing (A60 G), L/S 16, 50 ft. Applications: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance. Characteristics: Nonconductive, heat-sea
Catalog No. 50-110-8831