Learn More
Accuflow Systems Gore STA-PURE Pump Tubing, PCS Series, ePTFE/Silicone, 3/8 in
Supplier:  Accuflow Systems 9624173
Encompass_Preferred
GORE STA-PURE Pump Tubing (PCS Series) is a high-purity, composite peristaltic pump tubing designed for stable flow and extended run life in pharmaceutical and bioprocessing applications. Its ePTFE and platinum-cured silicone construction reduces spallation and resists ballooning or rupture at elevated pressures, making it suitable for long production cycles.
  • Composite construction of ePTFE and platinum-cured silicone.
  • Nominal inner diameter 3/8 in (9.5 mm).
  • Length 24 in (0.6 m) standard for PCS-24 items.
  • Suitable for peristaltic pump applications, compatible with I/P 73 pump heads.
  • Withstands operating pressures up to 100 psig for reliable performance.
  • Low spallation and extended pump life compared with silicone tubing.
Catalog No. NC1537059