Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Tubing, Non-Metallic, 1/8 in. x 0.062 in. x 11 in.

Supplier:  Waters Corp WAT07813

Encompass_Preferred

This non-metallic tubing, with precise dimensions of 1/8 in. x 0.062 in. x 11 in., is designed for various applications, particularly as part of service parts and kits.

  • Non-metallic construction.
  • Measures 1/8 in. diameter.
  • Features a 0.062 in. wall thickness.
  • Has a length of 11 in.

Catalog No. 50-793-722


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.