Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, Norprene A-60-G, L/S 13; 50 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0640413/DEL
Encompass
Applications: For applications requiring excellent chemical, heat, ozone, and ultraviolet (UV) light resistance Characteristics: Nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance, opaque black
Catalog No. NC9736758