Learn More
Microsolv Technology Corporation Tubing, PEEK, 0.055" ID, 1/16" OD, HPLC Grade, Black Stripe. 25 Meter Roll. 1 EA.
Supplier:  Microsolv Technology Corporation 4730555C
Encompass_Preferred
This HPLC Grade, PEEK tubing is extruded from virgin PEEK ( Victrex ) material without adding any additives. The red stripe is added only to the outside of the tubing leaving the interior all natural PEEK which eliminates leachables due to color. The internal and outer dimensional tolerances of this tubing is +/- 0.05mm. Color Code: Black Stripe for 0.055" ( 1.4mm ) ID. Pressure rating: 750 psi.
Catalog No. 50-227-1466