Learn More
Microsolv Technology Corporation Tubing, PEEK, Coiled, 0.010" ID, 1/16" OD. 1225mm L, General Grade, Blue, with Tubing Connectors. 1 EA.
Supplier:  Microsolv Technology Corporation 49307501B
Encompass_Preferred
These Flexible, Coiled PEEK Tubes will expand and contract which will save space in places such as behind an instrument and provide a Pulse Dampener Effect for Parabolic Flow Fluids which will result in a quieter base line and also reduce band broadening due to excess tubing lengths. This coil is 35mm wide when resting and 200mm when fully extended, with a total tubing length of 1225mm, end to end. Color Coded: Blue for 0.010" (0.25mm) ID
Catalog No. 50-227-1546