Learn More
Trajan Scientific Tubing PEEKSil Grey Medical 2/PK
SDP
Supplier:  Trajan Scientific 14010806000802
Encompass_Preferred
1/16 OD x 0 3 mm ID x 600 mm length PEEKsil tubing gray PK2
Catalog No. 50-004-94662