Learn More
Trajan Scientific Tubing PEEKSil Orange Medical 2/PK
SDP
Supplier:  Trajan Scientific 14020105500102
Encompass_Preferred
1/32 OD x 0 025 mm ID x 550 mm length PEEKsil tubing orange PK2
Catalog No. 50-004-93747