Learn More
Trajan Scientific Tubing PEEKSil Yellow Medical 2/PK
SDP
Supplier:  Trajan Scientific 14010600500602
Encompass_Preferred
1/16 OD x 0 175 mm ID x 50 mm length PEEKsil tubing yellow PK2
Catalog No. 50-004-93787