Learn More
Microsolv Technology Corporation Tubing, PFA, 0.030" ID, 1/16" OD, Low Pressure. 5 Meter Roll. 1 EA.
Supplier:  Microsolv Technology Corporation 4831030
Encompass_Preferred
This MicroSolv PFA ( PerFlourAlkoxy ) low pressure tubing is very similar to Teflon but is less permeable, more transparent and performs better in high temperature and pressure. Higher temperature than FEP are possible. This tubing is used in all low-pressure, laboratory applications including before and after an HPLC Pump such as with Mobile Phase Delivery and Solvent Waste Systems. Also known as Teflon FEP.Do not use this tubing with heat over 260⁰C. Internal and Outer Dimensional Tolerance: +/- 0.10mm Color: Natural PFA, Transparent, slight light blue tint ID: 0.030" ( 0.75 mm ) Pressure rating: 1075 psi.
Catalog No. 50-227-1516