Learn More
Microsolv Technology Corporation Tubing Pfa 1/16 X.020 5M
Supplier:  Microsolv Technology Corporation 4831020
Encompass_Preferred
Tubing, PFA, 0.020 ID, 1/16 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51603