Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, PFA, 1/2"" ID x 9/16"" OD; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0637108
Encompass
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. For the transfer of high-temperature or highly corrosive materials. Also good for high-purity work. Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent. Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -320 to 500¿F (-195 to 260¿C) Sterilize: Autoclave, ethylene oxide, or use dry heat.
Catalog No. 50-634-360