Learn More
Microsolv Technology Corporation Tubing Ptfe 1/8 X.1/16Th 5Mt
Supplier:  Microsolv Technology Corporation 4922565
Encompass_Preferred
Tubing, PTFE, 1/16 ID, 1/8 OD, Low Pressure. 5 Meter Roll. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51626