Learn More
Helix Medical Inc TUBING SIL .063 X .189 25FT
Supplier:  Helix Medical Inc 6082543
Encompass_Preferred
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.
Catalog No. NC9757991