Learn More
Helix Medical Inc TUBING SIL .188 X .313 50FT
Supplier:  Helix Medical Inc 6001122
Encompass_Preferred
Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.
Catalog No. NC0237169