Learn More
Helix Medical Inc TUBING SIL .313 X .501 25FT
Supplier:  Helix Medical Inc 60825163
Encompass_Preferred
Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing.
Catalog No. NC9036409