Learn More
New Age Industries Inc Platinum Cured Silicone Tubing | 5/8in ID | 50ft
SDP
Supplier:  New Age Industries Inc APST06250875L
Encompass_Preferred
Platinum Cured Silicone Tubing | 0.625in ID | 0.875in OD | 0.125in Wall | 50ft
Catalog No. NC1324989