Learn More
Accuflow Systems Masterflex L/S Spooled Silicone Tubing, Platinum-Cured, L/S 18, 400 ft
Supplier:  Accuflow Systems 9640318
Encompass_Preferred
Masterflex L/S 18 spooled silicone tubing for peristaltic pump applications. Supplied on a 400 ft (121.9 m) spool, the platinum-cured silicone formulation offers low extractables, flexibility, and chemical resistance for laboratory, pharmaceutical, and food-processing uses.
  • Platinum-cured silicone construction for biocompatibility and low extractables.
  • Size L/S 18, inner diameter 7.9 mm (0.31 in).
  • Supplied on 400 ft (121.9 m) spool for bulk applications.
  • Suitable for peristaltic pumps and fluid transfer in laboratory and process settings.
  • Good temperature resistance and flexibility to support long pump life.
Catalog No. NC2832347