Learn More
Accuflow Systems TUBING VINYL 1/2 x 1/16 50FT/PK
Supplier:  Accuflow Systems AGL00036
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 1/2 in. 12.7 mm Diameter Outer 5/8 in. 15.9 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15.2 m
Catalog No. 50-206-9017