Learn More
Med Vet International Dynarex Suction Connection Tubing w/Male Connector, Non-Conductive, 1/4" x 20' Tube, 20/Case
Supplier:  Med Vet International 32111
Encompass_Preferred
Dynarex Resp-O2™ Suction Connecting Tubing is designed to provide secure connections to both portable and in-wall suction sources. This non-conductive Tubing features female connectors on both ends and includes an optional male (straw) adapter for additional attachments. Designed to connect suction sources to suction catheters and yankauers Constructed from transparent material for clear visualization during suctioning Durable, crush-resistant tubing Single-use and disposable Not made with natural rubber latex or DEHP
Catalog No. 50-304-3217