Learn More
Med Vet International Dynarex Suction Connection Tubing w/ Male Connector, Non-Conductive,, 3/16" x 6' and 3/16" x 18" Tub, 50/Case
Supplier:  Med Vet International 32102
Encompass_Preferred
Dynarex Resp-O2™ Suction Connecting Tubing is designed to provide secure connections to portable and in-wall suction sources. This durable tubing offers the ultimate in versatility—featuring a female connector at each end and an optional rigid male (straw) adapter for attaching additional tubes. The high-quality, sterile tubing is disposable and designed for single use to promote optimal hygiene.  Dynarex Resp-O2™ Suction Connecting Tubing is designed to provide secure connections to both portable and in-wall suction sources. This non-conductive Tubing features female connectors on both ends and includes an optional male (straw) adapter for additional attachments.
Catalog No. 50-304-2894