Learn More
Medchemexpress LLC Tubulin cofactor A 1mg
SDP
Supplier:  Medchemexpress LLC HYP73565A1MG
Encompass_Preferred
The tubulin cofactor A protein is a key tubulin folding protein that initiates the tubulin folding pathway within the supercomplex (cofactors A to E) Cofactors A and D capture tubulin and stabilize it in a quasi-native state Tubulin cofactor A Protein Human (Tag Free) is the recombinant human-derived Tubulin cofactor A protein expressed by E coli with tag free
Catalog No. 50-001-80844