Learn More
TARGETMOL CHEMICALS INC TUBULIN INHIBITOR 8 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T132265MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tubulin inhibitor 8 is an inhibitor of tubulin and various cancer cell lines and inhibits tubulin polymerisation and K562 cell growth with an IC50 of 0.73 uM and 14 nM respectively. purity: 99%
Catalog No. 50-258-1368